3-Bromo-2-(methylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

23867-500MG 23867-1G 23867-5G 23867-25G
77324-492EA 157.59 USD
77324-492 77345-636 77360-816 77384-190
3-Bromo-2-(methylthio)phenylboronic acid ≥97%
3-Bromo-2-(methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂S
MW: 246.917 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28977124
CAS Number: 2121513-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR