4-Bromo-1-ethyl-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

23858-500MG 23858-1G 23858-5G 23858-25G
77324-490EA 87.55 USD
77324-490 77342-284 77360-814 77372-646
4-Bromo-1-ethyl-2-fluorobenzene ≥97%
4-Bromo-1-ethyl-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD16619427
CAS Number: 627463-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR