6-Fluoro-2,3'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

23752-500MG 23752-1G 23752-25G 23752-5G
77324-486EA 296.64 USD
77324-486 77348-354 77384-186 77372-644
6-Fluoro-2,3'-bipyridine ≥97%
6-Fluoro-2,3'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇FN₂
MW: 174.174 g/mol
Storage Temperature: Ambient
CAS Number: 1214335-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR