3-Chloro-4-fluoro-5-methoxybenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

23749-500MG 23749-1G 23749-5G
77338-882EA 528.39 USD
77338-882 77348-352 77372-642
3-Chloro-4-fluoro-5-methoxybenzoic acid methyl ester ≥97%
3-Chloro-4-fluoro-5-methoxybenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27934463
CAS Number: 950525-95-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR