4-Fluoro-3-methoxy-5-methylbenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

23748-500MG 23748-1G 23748-5G 23748-25G
77324-484EA 199.82 USD
77324-484 77348-350 77360-810 77384-184
4-Fluoro-3-methoxy-5-methylbenzoic acid methyl ester ≥97%
4-Fluoro-3-methoxy-5-methylbenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
MW: 198.191 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27934464
CAS Number: 950525-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR