3-Chloro-2-(methylsulfanyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

23741-500MG 23741-1G 23741-5G 23741-25G
77324-480EA 199.82 USD
77324-480 77348-348 77360-806 77384-180
3-Chloro-2-(methylsulfanyl)benzaldehyde ≥97%
3-Chloro-2-(methylsulfanyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇ClOS
MW: 186.659 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09859806
CAS Number: 1033574-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR