5-(Methylsulfanyl)thiophene-3-carboxylic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
23717-500MG
23717-1G
23717-5G
23717-25G
77324-474EA
218.36
USD
77324-474
77348-342
77360-800
77384-176
5-(Methylsulfanyl)thiophene-3-carboxylic acid ≥97%
Precaución: for Research Use Only
Formula:
C₆H₆O₂S₂ Storage Temperature: Ambient |
MDL Number:
MFCD20640762 CAS Number: 88511-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...