Benzenemethanol,2-fluoro-5-(2-fluoro-3-pyridinyl)- ≥95%

Supplier: AOBChem USA

Warning

23634-500MG 23634-1G 23634-5G
77335-492EA 356.38 USD
77335-492 77348-338 77372-636
Benzenemethanol,2-fluoro-5-(2-fluoro-3-pyridinyl)- ≥95%
Benzenemethanol,2-fluoro-5-(2-fluoro-3-pyridinyl)-

Precaución: for Research Use Only

Formula: C₁₂H₉F₂NO
MW: 221.203 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27943824
CAS Number: 1335512-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR