5-Bromo-4-methyl-3,4'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

23631-500MG 23631-1G 23631-5G
77324-470EA 312.09 USD
77324-470 77348-336 77372-634
5-Bromo-4-methyl-3,4'-bipyridine ≥97%
5-Bromo-4-methyl-3,4'-bipyridine

Precaución: for Research Use Only

Formula: C₁₁H₉BrN₂
MW: 249.107 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20488836
CAS Number: 351457-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR