3-Methoxy-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-Methoxy-2-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

23626-1G 23626-5G 23626-25G
77341-056EA 67.98 USD
77341-056 77360-796 77372-632
3-Methoxy-2-methylphenylboronic acid pinacol ester ≥97%
3-Methoxy-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22494163
CAS Number: 351456-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR