6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(4-(Trifluoromethyl)phenyl)nicotinaldehyde, 6-[4-(Trifluoromethyl)phenyl]-3-pyridinecarbaldehyde

23611-500MG 23611-1G 23611-5G 23611-25G
77324-464EA 129.78 USD
77324-464 77344-296 77360-790 77372-630
6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde ≥97%
6-[(4-Trifluoromethyl)phenyl]nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₈F₃NO
MW: 251.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115430
CAS Number: 356058-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR