5-Methyl-2-(4-methoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

23583-500MG 23583-1G 23583-25G 23583-5G
77324-462EA 289.43 USD
77324-462 77348-330 77384-168 77372-628
5-Methyl-2-(4-methoxyphenyl)pyridine ≥97%
5-Methyl-2-(4-methoxyphenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₃NO
MW: 199.248 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11040257
CAS Number: 25363-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR