2-(4-(1-methylethoxy)phenyl)pyridine-5-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

23576-500MG 23576-1G 23576-5G 23576-25G
77324-458EA 222.48 USD
77324-458 77348-326 77360-788 77384-166
2-(4-(1-methylethoxy)phenyl)pyridine-5-carboxaldehyde ≥97%
2-(4-(1-methylethoxy)phenyl)pyridine-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₂
MW: 241.285 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543661
CAS Number: 1213239-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR