5-Bromo-2-(2-fluoro-4-(trifluoromethyl)phenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

23569-500MG 23569-1G 23569-5G 23569-25G
77324-456EA 204.97 USD
77324-456 77348-322 77360-786 77384-164
5-Bromo-2-(2-fluoro-4-(trifluoromethyl)phenyl)pyridine ≥97%
5-Bromo-2-(2-fluoro-4-(trifluoromethyl)phenyl)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₆BrF₄N
MW: 320.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22418350
CAS Number: 1257875-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR