5-Bromo-2-[4-(trifluoromethoxy)phenyl]pyridine ≥97%

Supplier: AOBChem USA

Warning

23562-500MG 23562-1G 23562-5G 23562-25G
77324-452EA 154.5 USD
77324-452 77345-540 77360-782 77384-162
5-Bromo-2-[4-(trifluoromethoxy)phenyl]pyridine ≥97%
5-Bromo-2-[4-(trifluoromethoxy)phenyl]pyridine

Precaución: for Research Use Only

Formula: C₁₂H₇BrF₃NO
MW: 318.089 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22543659
CAS Number: 1257875-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR