6-Isopropoxy-4-methylnicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

23558-500MG 23558-1G 23558-5G 23558-25G
77324-450EA 251.32 USD
77324-450 77348-320 77360-780 77384-160
6-Isopropoxy-4-methylnicotinaldehyde ≥97%
6-Isopropoxy-4-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD22543658
CAS Number: 1289173-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR