3-(Hydroxymethyl)-4-methylphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Hydroxy-2-methylbenzyl alcohol

23540-500MG 23540-1G 23540-5G 23540-25G
77324-448EA 101.97 USD
77324-448 77342-980 77360-778 77372-620
3-(Hydroxymethyl)-4-methylphenol ≥97%
3-(Hydroxymethyl)-4-methylphenol

Precaución: for Research Use Only

Formula: C₈H₁₀O₂
MW: 138.164 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18397253
CAS Number: 1261454-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR