1-(5-Bromo-2-methoxy-benzyl)-pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

23437-500MG 23437-1G 23437-5G 23437-25G
77324-446EA 118.45 USD
77324-446 77343-546 77360-776 77372-616
1-(5-Bromo-2-methoxy-benzyl)-pyrrolidine ≥97%
1-(5-Bromo-2-methoxy-benzyl)-pyrrolidine

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO
MW: 270.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02105794
CAS Number: 864418-18-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR