5-Bromo-2-isopropoxy-4-methylpyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
23384-500MG
23384-1G
23384-5G
23384-25G
77324-436EA
146.26
USD
77324-436
77345-150
77360-768
77384-152
5-Bromo-2-isopropoxy-4-methylpyridine ≥97%
5-Bromo-2-isopropoxy-4-methylpyridine
Precaución: for Research Use Only
Formula:
C₉H₁₂BrNO MW: 230.102 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22056758 CAS Number: 1239611-37-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...