1-Bromo-4-chloro-2-fluoro-3-isobutoxybenzene ≥95%

Supplier: AOBChem USA

Warning

23366-500MG 23366-1G 23366-5G 23366-25G
77324-428EA 124.63 USD
77324-428 77344-042 77360-760 77372-610
1-Bromo-4-chloro-2-fluoro-3-isobutoxybenzene ≥95%
1-Bromo-4-chloro-2-fluoro-3-isobutoxybenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrClFO
Storage Temperature: Ambient
MDL Number: MFCD28402074
CAS Number: 1352718-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR