4,5-Dfluoro-2-methoxybenzoic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

23360-500MG 23360-1G 23360-5G 23360-25G
77324-426EA 187.46 USD
77324-426 77348-314 77360-758 77384-146
4,5-Dfluoro-2-methoxybenzoic acid ethyl ester ≥97%
4,5-Dfluoro-2-methoxybenzoic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11555008
CAS Number: 1256479-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR