5-Bromo-7-iodo-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

23302-500MG 23302-1G 23302-5G 23302-25G
77324-422EA 90.64 USD
77324-422 77342-416 77360-754 77372-608
5-Bromo-7-iodo-1H-indazole ≥97%
5-Bromo-7-iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrIN₂
MW: 322.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13177018
CAS Number: 953410-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR