4-Bromo-1,6-dimethyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

23278-500MG 23278-1G 23278-5G
77336-004EA 373.89 USD
77336-004 77348-312 77372-606
4-Bromo-1,6-dimethyl-1H-indazole ≥97%
4-Bromo-1,6-dimethyl-1H-indazole

Precaución: for Research Use Only

Formula: C₉H₉BrN₂
Storage Temperature: Ambient
MDL Number: MFCD12028621
CAS Number: 1159511-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR