2-Allyloxy-1-bromo-3-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

23126-500MG 23126-1G 23126-5G 23126-25G
77324-418EA 141.11 USD
77324-418 77344-778 77360-746 77384-140
2-Allyloxy-1-bromo-3-methylbenzene ≥95%
2-Allyloxy-1-bromo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD27930025
CAS Number: 854260-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR