4-(Benzyloxy)-3-chlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

23007-500MG 23007-1G 23007-5G 23007-25G
77324-414EA 149.35 USD
77324-414 77345-292 77360-742 77384-136
4-(Benzyloxy)-3-chlorobenzoic acid ≥97%
4-(Benzyloxy)-3-chlorobenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO₃
MW: 262.688 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203203
CAS Number: 106931-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR