Methyl 6-bromo-2,3-dichlorobenzoate ≥95%

Supplier: AOBChem USA

Warning

22997-500MG 22997-1G 22997-5G
77335-526EA 357.41 USD
77335-526 77348-306 77372-598
Methyl 6-bromo-2,3-dichlorobenzoate ≥95%
Methyl 6-bromo-2,3-dichlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O₂
MW: 283.934 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11976596
CAS Number: 1257856-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR