2-Fluoro-4-methylpyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

22839-500MG 22839-1G 22839-5G
77336-166EA 380.07 USD
77336-166 77348-304 77372-596
2-Fluoro-4-methylpyridine-3-carboxylic acid ≥97%
2-Fluoro-4-methylpyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13189141
CAS Number: 1060804-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR