2-Fluoro-6-iodo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22830-500MG 22830-1G 22830-5G 22830-25G
77324-412EA 198.79 USD
77324-412 77348-302 77360-740 77384-134
2-Fluoro-6-iodo-3-methylbenzaldehyde ≥97%
2-Fluoro-6-iodo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD11847123
CAS Number: 1260839-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR