4-Fluoro-3,5-dimethylphenol ≥95%

Supplier: AOBChem USA

Warning

22812-500MG 22812-1G 22812-5G
77339-660EA 611.82 USD
77339-660 77348-300 77372-594
4-Fluoro-3,5-dimethylphenol ≥95%
4-Fluoro-3,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉FO
MW: 140.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405442
CAS Number: 1043450-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR