7-Bromo-4-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22795-500MG 22795-1G 22795-25G 22795-5G
77324-408EA 287.37 USD
77324-408 77348-296 77384-132 77372-590
7-Bromo-4-methyl-1H-indazole ≥97%
7-Bromo-4-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17168274
CAS Number: 1257535-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR