7-Bromo-6-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

22793-500MG 22793-1G 22793-5G 22793-25G
77324-406EA 150.38 USD
77324-406 77345-382 77360-738 77384-130
7-Bromo-6-methyl-1H-indazole ≥97%
7-Bromo-6-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
Storage Temperature: Ambient
MDL Number: MFCD17168273
CAS Number: 1257535-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR