7-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95%

Supplier: AOBChem USA

Warning

22792-500MG 22792-1G 22792-5G 22792-25G
77324-404EA 150.38 USD
77324-404 77345-332 77360-736 77384-128
7-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole ≥95%
7-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrN₂O
MW: 281.148 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22380250
CAS Number: 1158680-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR