7-Fluoro-3-iodo-6-methoxy-1H-indazole ≥95%
Supplier: AOBChem USA
|
Warning
|
22756-500MG
22756-1G
22756-5G
22756-25G
77324-400EA
223.51
USD
77324-400
77348-294
77360-732
77384-124
7-Fluoro-3-iodo-6-methoxy-1H-indazole ≥95%
7-Fluoro-3-iodo-6-methoxy-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₆FIN₂O Storage Temperature: Ambient |
MDL Number:
MFCD32706996 CAS Number: 2484889-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...