2,4-dimethoxy-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

22754-500MG 22754-1G 22754-5G
77339-836EA 656.11 USD
77339-836 77348-292 77372-588
2,4-dimethoxy-3-fluorobenzaldehyde ≥97%
2,4-dimethoxy-3-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉FO₃
MW: 184.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24685833
CAS Number: 502483-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR