4-Iodo-3-methoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

22709-500MG 22709-1G 22709-5G 22709-25G
77324-398EA 82.4 USD
77324-398 77342-140 77360-728 77372-584
4-Iodo-3-methoxybenzaldehyde ≥95%
4-Iodo-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11847312
CAS Number: 121404-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR