4-Bromo-2,5-dichlorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22612-1G 22612-5G 22612-25G
77340-606EA 60.77 USD
77340-606 77360-722 77372-576
4-Bromo-2,5-dichlorobenzoic acid ≥97%
4-Bromo-2,5-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
MW: 269.91 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08444534
CAS Number: 885532-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR