2-Trimethylsilylanisole ≥97%

Supplier: AOBChem USA

Warning

22436-500MG 22436-1G 22436-5G 22436-25G
77324-392EA 183.34 USD
77324-392 77347-110 77360-718 77384-118
2-Trimethylsilylanisole ≥97%
2-Trimethylsilylanisole

Precaución: for Research Use Only

Formula: C₁₀H₁₆OSi
Storage Temperature: Ambient
MDL Number: MFCD09039086
CAS Number: 704-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR