(3-Bromo-5-chlorophenyl)(pyrrolidin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
22364-500MG
22364-1G
22364-5G
22364-25G
77324-388EA
211.15
USD
77324-388
77348-274
77360-714
77384-114
(3-Bromo-5-chlorophenyl)(pyrrolidin-1-yl)methanone ≥97%
(3-Bromo-5-chlorophenyl)(pyrrolidin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₁H₁₁BrClNO MW: 288.568 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD32878176 CAS Number: 1326987-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...