1-[(4-Bromo-2,6-difluorophenyl)carbonyl]pyrrolidine ≥97%

Supplier: AOBChem USA

Warning

22354-1G 22354-5G 22354-25G
77341-544EA 92.7 USD
77341-544 77360-710 77372-564
1-[(4-Bromo-2,6-difluorophenyl)carbonyl]pyrrolidine ≥97%
1-[(4-Bromo-2,6-difluorophenyl)carbonyl]pyrrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrF₂NO
MW: 290.104 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526376
CAS Number: 1351668-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR