3-Cyclopropylpyridine ≥97%

Supplier: AOBChem USA

Warning

22312-500MG 22312-1G 22312-25G 22312-5G
77324-384EA 299.73 USD
77324-384 77348-272 77384-110 77372-562
3-Cyclopropylpyridine ≥97%
3-Cyclopropylpyridine

Precaución: for Research Use Only

Formula: C₈H₉N
Storage Temperature: Ambient
MDL Number: MFCD17169885
CAS Number: 188918-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR