1-Chloro-3,5-dicyclopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

22307-500MG 22307-1G 22307-5G
77339-284EA 572.68 USD
77339-284 77348-270 77372-560
1-Chloro-3,5-dicyclopropylbenzene ≥97%
1-Chloro-3,5-dicyclopropylbenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₃Cl
Storage Temperature: Ambient
MDL Number: MFCD28365397
CAS Number: 1202644-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR