2-Fluoro-6-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: [2-Fluoro-6-(trifluoromethoxy)phenyl]boronic acid

22283-500MG 22283-1G 22283-5G 22283-25G
77324-382EA 276.04 USD
77324-382 77348-268 77360-708 77384-108
2-Fluoro-6-(trifluoromethoxy)phenylboronic acid ≥97%
2-Fluoro-6-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₄O₃
MW: 223.917 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22551457
CAS Number: 1261214-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR