Ethyl 5-chloro-2-ethoxypyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

22120-500MG 22120-1G 22120-5G 22120-25G
77324-376EA 172.01 USD
77324-376 77346-412 77360-702 77384-102
Ethyl 5-chloro-2-ethoxypyridine-3-carboxylate ≥97%
Ethyl 5-chloro-2-ethoxypyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClNO₃
MW: 229.66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105410
CAS Number: 1310416-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR