2,3-Dichloro-N-methyl-4-pyridinecarboxamide ≥97%

Supplier: AOBChem USA

Warning

22091-500MG 22091-1G 22091-5G 22091-25G
77324-370EA 118.45 USD
77324-370 77343-544 77360-696 77372-556
2,3-Dichloro-N-methyl-4-pyridinecarboxamide ≥97%
2,3-Dichloro-N-methyl-4-pyridinecarboxamide

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂O
MW: 205.041 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03411652
CAS Number: 329794-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR