5-Chloro-2-hydroxy-3-(trifluoromethyl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

22052-500MG 22052-1G 22052-5G
77338-316EA 476.89 USD
77338-316 77348-260 77372-554
5-Chloro-2-hydroxy-3-(trifluoromethyl)benzaldehyde ≥95%
5-Chloro-2-hydroxy-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MW: 224.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28647456
CAS Number: 1235783-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR