3-(4-Morpholinylcarbonyl)-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

22021-500MG 22021-1G 22021-5G 22021-25G
77324-366EA 166.86 USD
77324-366 77346-188 77360-690 77384-096
3-(4-Morpholinylcarbonyl)-5-(trifluoromethyl)phenylboronic acid ≥97%

Precaución: for Research Use Only

Formula: C₁₂H₁₃BF₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD27665150
CAS Number: 1264616-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR