3-Chloro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%

Supplier: AOBChem USA

Warning

22010-1G 22010-5G 22010-25G
77340-944EA 64.89 USD
77340-944 77360-684 77372-544
3-Chloro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide ≥97%
3-Chloro-N-methoxy-N-methyl-5-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₀H₉ClF₃NO₂
MW: 267.632 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755774
CAS Number: 1076229-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR