3-Chloro-5-(trifluoromethyl)benzoic acid ethyl ester ≥97%
Supplier: AOBChem USA
|
Warning
|
22008-500MG
22008-1G
22008-5G
22008-25G
77324-360EA
87.55
USD
77324-360
77342-282
77360-682
77372-542
3-Chloro-5-(trifluoromethyl)benzoic acid ethyl ester ≥97%
3-Chloro-5-(trifluoromethyl)benzoic acid ethyl ester
Precaución: for Research Use Only
Formula:
C₁₀H₈ClF₃O₂ Storage Temperature: Ambient |
MDL Number:
MFCD14698205 CAS Number: 1214366-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...