Ethyl 2-fluoro-3-methylpyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

21997-500MG 21997-1G 21997-5G
77336-448EA 388.31 USD
77336-448 77348-252 77372-538
Ethyl 2-fluoro-3-methylpyridine-4-carboxylate ≥97%
Ethyl 2-fluoro-3-methylpyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610956
CAS Number: 1227508-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR