4-Bromo-4'-isobutoxybiphenyl ≥97%

Supplier: AOBChem USA

Warning

21967-1G 21967-5G 21967-25G
77341-300EA 82.4 USD
77341-300 77360-678 77372-536
4-Bromo-4'-isobutoxybiphenyl ≥97%
4-Bromo-4'-isobutoxybiphenyl

Precaución: for Research Use Only

Formula: C₁₆H₁₇BrO
MW: 305.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19105398
CAS Number: 1310416-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR